Proteintech
Manufacturer Code:265451AP150UL
Catalog # 50-173-5370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZDHHC1 Fusion Protein Ag24028 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf1 DHHC 1 HSU90653 ZDHHC1 Zinc finger protein 377 ZNF377; DHHC domain-containing cysteine-rich protein 1; DHHC-1; DHHC-domain-containing cysteine-rich protein; Palmitoyltransferase ZDHHC1; Zinc finger DHHC domain-containing protein 1; Zinc finger protein 377; zinc finger, DHHC domain containing 1; zinc finger, DHHC-type containing 1
Gene Aliases: C16orf1; HSU90653; ZDHHC1; ZNF377
UniProt ID: (Human) Q8WTX9
Entrez Gene ID: (Human) 29800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.