Novus Biologicals
Manufacturer Code:NBP288611
Catalog # NB036233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human ZCCHC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp666B142 DKFZp686C11112 DKFZp686F119 DKFZp686I1269 EC 2.7.7.52 FLJ13409 HS2 KIAA1711 PAP associated domain containing 6 PAPD6 terminal uridylyltransferase 7 TUT7 TUTase 7 Zinc finger CCHC domain-containing protein 6 zinc finger CCHC domain containing 6; PAP associated domain containing 6; Terminal uridylyltransferase 7; TUTase 7; Zinc finger CCHC domain-containing protein 6; zinc finger, CCHC domain containing 6
Gene Aliases: HS2; KIAA1711; PAPD6; TUT7; ZCCHC6
UniProt ID: (Human) Q5VYS8
Entrez Gene ID: (Human) 79670
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.