Novus Biologicals
Manufacturer Code:NBP181888
Catalog # NBP181888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QDPATFHLLAQNLICNETNQSGQFEKALADPNQDEESVTDMMDNLPDLITQCIQLDKKHSDRPELLQSETQLPLLASLIQHQALFSPTDPPPKKGPIQLREGQLPLTPAKRARQQETQLCLYCSQSGHFTRDCLAKRSRAPATTNNTAH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ46608 mammalian retrotransposon-derived protein 4 MAR3 Mar4 Mart4 zinc finger CCHC domain-containing protein 16 zinc finger CCHC domain containing 16; Mammalian retrotransposon-derived protein 4; Retrotransposon Gag-like protein 4; Zinc finger CCHC domain-containing protein 16; zinc finger, CCHC domain containing 16
Gene Aliases: MAR3; Mar4; MART4; RTL4; ZCCHC16
UniProt ID: (Human) Q6ZR62
Entrez Gene ID: (Human) 340595
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.