Invitrogen
Manufacturer Code:MA524539
Catalog # PIMA524539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G11 |
Immunogen | ZCCHC13 (NP&lowbar976048.1 1 a.a. &bsim 166 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ZCCHC13; Zinc finger CCHC domain-containing protein 13; zinc finger, CCHC domain containing 13
Gene Aliases: CNBP2; ZCCHC13; ZNF9L
UniProt ID: (Human) Q8WW36
Entrez Gene ID: (Human) 389874
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.