Abnova
Manufacturer Code:H00170261K
Catalog # 89-206-643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ZCCHC12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ZCCHC12 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ16123 SIZN SIZN1; paraneoplastic Ma antigen family member 7A; Smad-interacting zinc finger protein 1; Zinc finger CCHC domain-containing protein 12; zinc finger, CCHC domain containing 12
Gene Aliases: PNMA7A; SIZN; SIZN1; ZCCHC12
UniProt ID: (Human) Q6PEW1
Entrez Gene ID: (Human) 170261
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.