Invitrogen
Manufacturer Code:PA555604
Catalog # PIPA555604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZCCHC11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAP associated domain containing 3; Terminal uridylyltransferase 4; TUTase 4; zinc finger CCHC domain containing 11 RP23-406B13.1 6030404K05Rik 9230115F04Rik KIAA0191 PPAPD3 mKIAA0191 TUTase 4 terminal uridylyltransferase 4 zinc finger CCHC domain-containing protein 11; Zinc finger CCHC domain-containing protein 11; zinc finger, CCHC domain containing 11
Gene Aliases: KIAA0191; PAPD3; TUT4; ZCCHC11
UniProt ID: (Human) Q5TAX3
Entrez Gene ID: (Human) 23318
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.