Abnova
Manufacturer Code:PAB24626
Catalog # 89-126-341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Zcchc11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Zcchc11.
For Research Use Only
Protein Aliases: 6030404K05Rik; 9230115F04Rik; KIAA0191; mKIAA0191; PPAPD3; Terminal uridylyltransferase 4; TUTase 4; Zinc finger CCHC domain-containing protein 11
Gene Aliases: 6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PPAPD3; Tut4; Zcchc11
UniProt ID: (Mouse) B2RX14
Entrez Gene ID: (Mouse) 230594
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.