Novus Biologicals
Manufacturer Code:NBP177184
Catalog # NBP177184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human ZC3HAV1. The immunogen is located within amino acids 680 - 730 of ZC3HAV1. Amino Acid Squence: QMKRGPDHQPAKTS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C7orf39 chromosome 7 open reading frame 39 MGC14289 zinc finger CCCH-type antiviral protein 1-like zinc finger CCCH-type antiviral 1-like; Zinc finger CCCH-type antiviral protein 1-like; zinc finger CCCH-type, antiviral 1-like
Gene Aliases: C7orf39; ZC3HAV1L
UniProt ID: (Human) Q96H79
Entrez Gene ID: (Human) 92092
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.