Proteintech
Manufacturer Code:168201AP150UL
Catalog # 50-173-1199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZC3HAV1 Fusion Protein Ag10364 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; CCCH-type zinc finger antiviral protein (Zap); FLB6421 ZAP ZC3H2 ZC3HAV1 ZC3HDC2 Zinc finger antiviral protein; Inactive Poly [ADP-ribose] polymerase 13; PARP13; rZAP; ZAP; Zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; Zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type, antiviral 1
Gene Aliases: 1200014N16Rik; 2900058M19Rik; 9130009D18Rik; 9830115L13Rik; ARTD13; D6Bwg1452e; FLB6421; PARP13; PRO1677; ZAP; ZC3H2; ZC3HAV1; ZC3HDC2
UniProt ID: (Human) Q7Z2W4, (Mouse) Q3UPF5
Entrez Gene ID: (Human) 56829, (Rat) 252832, (Mouse) 78781
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.