Abnova
Manufacturer Code:H00056829K
Catalog # 89-201-914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ZC3HAV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ZC3HAV1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; DKFZp686F2052 DKFZp686H1869 DKFZp686O19171 FLB6421 FLJ13288 MGC48898 ZAP ZC3H2 ZC3HDC2; Inactive Poly [ADP-ribose] polymerase 13; PARP13; ZAP; Zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; Zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type, antiviral 1
Gene Aliases: ARTD13; FLB6421; PARP13; PRO1677; ZAP; ZC3H2; ZC3HAV1; ZC3HDC2
UniProt ID: (Human) Q7Z2W4
Entrez Gene ID: (Human) 56829
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.