Novus Biologicals
Manufacturer Code:NBP274908V
Catalog # NBP274908V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H5 |
Immunogen | Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Fliz1 ZC3HDC8 zinc finger CCCH domain-containing protein 8 zinc finger CCCH type domain containing 8 zinc finger CCCH-type containing 8 zinc finger CCCH-type domain containing 8; Zinc finger CCCH domain-containing protein 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type domain containing 8
Gene Aliases: Fliz1; ZC3H8; ZC3HDC8
UniProt ID: (Human) Q8N5P1
Entrez Gene ID: (Human) 84524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.