Invitrogen
Manufacturer Code:PA575525
Catalog # PIPA575525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 133-178 of Human ZC3H8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; Fetal liver zinc finger protein 1; Zinc finger CCCH domain-containing protein 8; Zinc finger CCCH domain-containing protein 8 ZC3HDC8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type domain containing 8
Gene Aliases: AU020882; E130108N08Rik; Fliz1; ZC3H8; ZC3HDC8
UniProt ID: (Human) Q8N5P1, (Mouse) Q9JJ48
Entrez Gene ID: (Human) 84524, (Rat) 311414, (Mouse) 57432
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.