Invitrogen
Manufacturer Code:PA598505
Catalog # PIPA598505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Zinc finger CCCH domain-containing protein 14 protein (176-306AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mammalian suppressor of tau pathology-2; MSUT-2; nuclear protein UKp68; Renal carcinoma antigen NY-REN-37; Zinc finger CCCH domain-containing protein 14
Gene Aliases: MSUT-2; NY-REN-37; SUT2; UKp68; ZC3H14
UniProt ID: (Human) Q6PJT7
Entrez Gene ID: (Human) 79882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.