Invitrogen
Manufacturer Code:PA587844
Catalog # PIPA587844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-306 of human ZC3H14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mammalian suppressor of tau pathology-2; MSUT-2; nuclear protein UKp68; Nuclear protein UKp83/UKp68; Renal carcinoma antigen NY-REN-37; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH-type containing 14
Gene Aliases: 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; MSUT-2; Npuk68; NY-REN-37; SUT2; UKp68; ZC3H14
UniProt ID: (Human) Q6PJT7, (Rat) Q7TMD5, (Mouse) Q8BJ05
Entrez Gene ID: (Human) 79882, (Rat) 192359, (Mouse) 75553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.