Novus Biologicals
Manufacturer Code:NBP156914
Catalog # NBP156914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ZC3H14(zinc finger CCCH-type containing 14) The peptide sequence was selected from the N terminal of ZC3H14. Peptide sequence KFPSPPLPIFLPPEPVDLGSITSSSCSLNELDNISHLLRKISADINEIKG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ11806 Mammalian suppressor of tau pathology-2 MGC26892 MSUT-2 nuclear protein UKp68 NY-REN-37 Renal carcinoma antigen NY-REN-37 SUT2 UKp68 zinc finger CCCH domain-containing protein 14 zinc finger CCCH-type containing 14; Mammalian suppressor of tau pathology-2; MSUT-2; nuclear protein UKp68; Renal carcinoma antigen NY-REN-37; Zinc finger CCCH domain-containing protein 14
Gene Aliases: MSUT-2; NY-REN-37; SUT2; UKp68; ZC3H14
UniProt ID: (Human) Q6PJT7
Entrez Gene ID: (Human) 79882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.