Invitrogen
Manufacturer Code:PA557865
Catalog # PIPA557865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZC3H12D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCP-induced protein 4; p34; Probable ribonuclease ZC3H12D; RP1-281H8.5 C6orf95 MCPIP4 TFL dJ281H8.1 p34 MCP-induced protein 4 Zinc finger CCCH domain-containing protein 12D probable ribonuclease ZC3H12D transformed follicular lymphoma tumor suppressor TFL; Transformed follicular lymphoma; tumor suppressor TFL; Zinc finger CCCH domain-containing protein 12D
Gene Aliases: C6orf95; dJ281H8.1; MCPIP4; p34; TFL; ZC3H12D
UniProt ID: (Human) A2A288
Entrez Gene ID: (Human) 340152
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.