Novus Biologicals
Manufacturer Code:NBP281878
Catalog # NB017189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZC3H12D antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZC3H12D. The immunogen is located within amino acids 280 - 330 of ZC3H12D. Amino Acid Squence: TGARPGAGAEEQRPPRAPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf95 FLJ46041 MCP-induced protein 4 p34 TFL transformed follicular lymphoma tumor suppressor TFL zinc finger CCCH-type containing 12D; MCP-induced protein 4; p34; Probable ribonuclease ZC3H12D; Transformed follicular lymphoma; tumor suppressor TFL; Zinc finger CCCH domain-containing protein 12D
Gene Aliases: C6orf95; dJ281H8.1; MCPIP4; p34; TFL; ZC3H12D
UniProt ID: (Human) A2A288
Entrez Gene ID: (Human) 340152
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.