Proteintech
Manufacturer Code:250091AP150UL
Catalog # 50-173-4667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZC3H12A Fusion Protein Ag13877 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endoribonuclease ZC3H12A; MCP induced protein 1 MCPIP MCPIP1 Ribonuclease ZC3H12A RP3 423B22.1 ZC3H12A; MCP-1 treatment-induced protein; MCP-induced protein 1; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; ribonuclease ZC3H12A; zc3h12a; Zinc finger CCCH domain-containing protein 12A
Gene Aliases: BC036563; dJ423B22.1; MCPIP; MCPIP1; ZC3H12A
UniProt ID: (Human) Q5D1E8, (Mouse) Q5D1E7
Entrez Gene ID: (Human) 80149, (Mouse) 230738
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.