Invitrogen
Manufacturer Code:PA539539
Catalog # PIPA539539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ZC11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Zinc finger CCCH domain-containing protein 11A; Zinc finger CCCH domain-containing protein 11A KIAA0663 ZC3HDC11A; zinc finger CCCH-type domain containing 11A
Gene Aliases: 1110003F06Rik; 5730454B08Rik; G630041M05Rik; KIAA0663; mKIAA0663; ZC3H11A; ZC3HDC11A; Zc3hh11a
UniProt ID: (Human) O75152, (Mouse) Q6NZF1
Entrez Gene ID: (Human) 9877, (Mouse) 70579
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.