Novus Biologicals
Manufacturer Code:NBP182973
Catalog # NBP182973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ETMREKHMQKQQEREKSVLTPLRGDVASCNTQVAEKPVLTAVPGITRHLTKRLPTKSSQKVEVETSGIGDSLLNVKCAAQTLEKRGKAKPKVNVKPSVVKVV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686D03108 DKFZp686F14109 DKFZp781G2455 EC 4.1.1.31 EC 4.2.1.11 KIAA0663zinc finger CCCH-type domain containing 11A ZC3HDC11A zinc finger CCCH domain-containing protein 11A zinc finger CCCH-type containing 11A; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH-type domain containing 11A
Gene Aliases: KIAA0663; ZC3H11A; ZC3HDC11A
UniProt ID: (Human) O75152
Entrez Gene ID: (Human) 9877
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.