Novus Biologicals
Manufacturer Code:NB10074650
Catalog # NB10074650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human zinc finger CCCH-type containing 11A using the numbering given in entry NP_055642.3 (GeneID 9877). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686D03108 DKFZp686F14109 DKFZp781G2455 EC 4.1.1.31 EC 4.2.1.11 KIAA0663zinc finger CCCH-type domain containing 11A ZC3HDC11A zinc finger CCCH domain-containing protein 11A zinc finger CCCH-type containing 11A; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH-type domain containing 11A
Gene Aliases: KIAA0663; ZC3H11A; ZC3HDC11A
UniProt ID: (Human) O75152
Entrez Gene ID: (Human) 9877
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.