Invitrogen
Manufacturer Code:PA571311
Catalog # PIPA571311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human ZBTB8B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: putative zinc finger and BTB domain-containing protein 8B; zinc finger and BTB domain containing 8; Zinc finger and BTB domain-containing protein 8B
Gene Aliases: AV220464; BC023839; BOZF1; RGD1562327; Zbtb8; ZBTB8B; Zbtb8l; ZNF916B
UniProt ID: (Human) Q8NAP8, (Mouse) Q8CII0
Entrez Gene ID: (Human) 728116, (Horse) 100630269, (Dog) 487306, (Rabbit) 100341984, (Guinea pig) 100728711, (Mouse) 215627, (Rat) 500553
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.