Novus Biologicals
Manufacturer Code:NBP2837890.1ML
Catalog # NB031718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human ZBTB8B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp547H154 RP1-27O5.1 zinc finger and BTB domain containing 8B zinc finger and BTB domain-containing protein 8B ZNF916B; putative zinc finger and BTB domain-containing protein 8B; Zinc finger and BTB domain-containing protein 8B
Gene Aliases: ZBTB8B; ZNF916B
UniProt ID: (Human) Q8NAP8
Entrez Gene ID: (Human) 728116
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.