Invitrogen
Manufacturer Code:PA568525
Catalog # PIPA568525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the c terminal region of human ZBTB8A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOZ-F1; BOZF1 ZBTB8 ZNF916A BOZ-F1 BTB/POZ and zinc-finger domain-containing factor BTB/POZ and zinc-finger domains factor on chromosome 1 zinc finger and BTB domain containing 8 zinc finger and BTB domain-containing protein 8A; BTB/POZ and zinc-finger domain-containing factor; BTB/POZ and zinc-finger domains factor on chromosome 1; zinc finger and BTB domain containing 8; Zinc finger and BTB domain-containing protein 8A
Gene Aliases: 2410081M15Rik; AI480941; BOZF1; ZBTB8; ZBTB8A; ZNF916A
UniProt ID: (Human) Q96BR9, (Rat) B1WBU4, (Mouse) Q9CWH1
Entrez Gene ID: (Rabbit) 100353816, (Horse) 100055796, (Human) 653121, (Dog) 487304, (Guinea pig) 100726759, (Rat) 313049, (Mouse) 73680
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.