Invitrogen
Manufacturer Code:PA568994
Catalog # PIPA568994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat ZBTB44 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB (POZ) domain containing 15; BTB/POZ domain-containing protein 15; Zinc finger and BTB domain-containing protein 44; Zinc finger and BTB domain-containing protein 44 BTB/POZ domain-containing protein 15 Zinc finger protein 851 ZBTB44 BTBD15 ZNF851; Zinc finger protein 851
Gene Aliases: 6030404E16Rik; BTBD15; E230011F09; HSPC063; RGD1308984; ZBTB44; ZNF851
UniProt ID: (Human) Q8NCP5, (Rat) Q3SWU4, (Mouse) Q8R0A2
Entrez Gene ID: (Dog) 489277, (Rabbit) 100338293, (Horse) 100064712, (Human) 29068, (Guinea pig) 100713204, (Rat) 363035, (Mouse) 235132
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.