Proteintech
Manufacturer Code:254211AP150UL
Catalog # 50-173-4900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZBTB44 Fusion Protein Ag22299 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB (POZ) domain containing 15; BTB/POZ domain-containing protein 15; BTBD15 HSPC063 ZBTB44 ZNF851; Zinc finger and BTB domain-containing protein 44; Zinc finger protein 851
Gene Aliases: BTBD15; HSPC063; ZBTB44; ZNF851
UniProt ID: (Human) Q8NCP5
Entrez Gene ID: (Human) 29068
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.