Abnova
Manufacturer Code:H00029068K
Catalog # 89-209-368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ZBTB44 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ZBTB44 peptide using ARM Technology.
For Research Use Only
Protein Aliases: BTB (POZ) domain containing 15; BTB/POZ domain-containing protein 15; BTBD15 HSPC063 MGC26123 MGC57431 MGC60348 MGC88058 ZNF851; Zinc finger and BTB domain-containing protein 44; Zinc finger protein 851
Gene Aliases: BTBD15; HSPC063; ZBTB44; ZNF851
UniProt ID: (Human) Q8NCP5
Entrez Gene ID: (Human) 29068
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.