Invitrogen
Manufacturer Code:PA5101960
Catalog # PIPA5101960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ZBTB40 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC059177 C230087D24 Gm571 KIAA0478 mKIAA0478 RGD1309866 ZBTB40 zinc finger and BTB domain containing 40 zinc finger and BTB domain-containing protein 40 ZNF923; Zinc finger and BTB domain-containing protein 40
Gene Aliases: BC059177; C230087D24; Gm571; KIAA0478; mKIAA0478; ZBTB40; ZNF923
UniProt ID: (Human) Q9NUA8
Entrez Gene ID: (Human) 9923, (Mouse) 230848
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.