Novus Biologicals
Manufacturer Code:NBP288605
Catalog # NB036227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of ZBTB38 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CIBZ FLJ22332 FLJ31131 FLJ35036 zinc finger and BTB domain containing 38 zinc finger and BTB domain-containing protein 38 ZNF921; protein phosphatase 1, regulatory subunit 171; ZBTB38; Zinc finger and BTB domain-containing protein 38
Gene Aliases: CIBZ; PPP1R171; ZBTB38; ZNF921
UniProt ID: (Human) Q8NAP3
Entrez Gene ID: (Human) 253461
Molecular Function: DNA binding protein centromere DNA-binding protein nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.