Proteintech
Manufacturer Code:256331AP150UL
Catalog # 50-173-5001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZBTB22 Fusion Protein Ag22439 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BING1 fru fruitless Protein BING1 ZBTB22 ZBTB22A Zinc finger protein 297 ZNF297 ZNF297A; Protein BING1; Zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; Zinc finger protein 297
Gene Aliases: 1110008J20Rik; AI265210; AI415166; BING1; fru; fruitless; ZBTB22; ZBTB22A; Zfp297; ZNF297; ZNF297A
UniProt ID: (Human) O15209, (Mouse) Q9Z0G7
Entrez Gene ID: (Human) 9278, (Mouse) 81630
Molecular Function: actin family cytoskeletal protein cytoskeletal protein hydrolase non-motor actin binding protein protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.