Abnova
Manufacturer Code:H00009278K
Catalog # 89-204-508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ZBTB22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ZBTB22 peptide using ARM Technology.
For Research Use Only
Protein Aliases: BING1 ZBTB22A ZNF297 ZNF297A fru fruitless; Protein BING1; Zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; Zinc finger protein 297
Gene Aliases: BING1; fru; fruitless; ZBTB22; ZBTB22A; ZNF297; ZNF297A
UniProt ID: (Human) O15209
Entrez Gene ID: (Human) 9278
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.