Invitrogen
Manufacturer Code:PA531896
Catalog # PIPA531896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 668 of Human ZBTB20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain zinc finger factor HOF; BTB/POZ zinc finger protein DPZF; dendritic cell-derived BTB/POZ zinc finger; Dendritic-derived BTB/POZ zinc finger protein; DPZF HOF ODA-8S ZNF288 BTB/POZ zinc finger protein DPZF dendritic cell-derived BTB/POZ zinc finger dendritic-derived BTB/POZ zinc finger protein zinc finger and BTB domain-containing protein 20 zinc finger protein 288; POZ/zinc finger transcription factor ODA-8; Zinc finger and BTB domain-containing protein 20; Zinc finger protein 288
Gene Aliases: 1300017A20Rik; 7330412A13Rik; A930017C21Rik; D16Wsu73e; DPZF; HOF; ODA-8S; Oda8; PRIMS; ZBTB20; Zfp288; ZNF288
UniProt ID: (Human) Q9HC78, (Mouse) Q8K0L9
Entrez Gene ID: (Human) 26137, (Mouse) 56490
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.