Novus Biologicals
Manufacturer Code:NBP30437020UL
Catalog # NB095892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human ZBTB20 (NP_056457.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ zinc finger protein DPZF; BTB/POZ zinc finger protein DPZF dendritic cell-derived BTB/POZ zinc finger Dendritic-derived BTB/POZ zinc finger protein DPZFZinc finger protein 288ZNF288DKFZp566F123 FLJ26458 HOF ODA-8S zinc finger and BTB domain containing 20 zinc finger and BTB domain-containing protein 20; dendritic cell-derived BTB/POZ zinc finger; Dendritic-derived BTB/POZ zinc finger protein; Zinc finger and BTB domain-containing protein 20; Zinc finger protein 288
Gene Aliases: DPZF; HOF; ODA-8S; PRIMS; ZBTB20; ZNF288
UniProt ID: (Human) Q9HC78
Entrez Gene ID: (Human) 26137
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.