Novus Biologicals
Manufacturer Code:NBP224660SS
Catalog # NBP224660SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 450 to 500 of human ZBTB2 was used as immunogen for the antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA351K16.2 zinc finger and BTB domain containing 2 zinc finger and BTB domain-containing protein 2 ZNF437KIAA1483; Zinc finger and BTB domain-containing protein 2
Gene Aliases: KIAA1483; ZBTB2; ZNF437
UniProt ID: (Human) Q8N680
Entrez Gene ID: (Human) 57621
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.