OriGene
Manufacturer Code:CF811343
Catalog # 50-167-0603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7D8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 112-457 of human ZBTB17 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Miz-1; MIZ-1 ZNF151 ZNF60 pHZ-67; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; Zinc finger and BTB domain-containing protein 17; Zinc finger protein 151; zinc finger protein 151 (pHZ-67); Zinc finger protein 60
Gene Aliases: MIZ-1; MIZ1; pHZ-67; ZBTB17; ZNF151; ZNF60
UniProt ID: (Human) Q13105
Entrez Gene ID: (Human) 7709
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.