Invitrogen
Manufacturer Code:PA5103320
Catalog # PIPA5103320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ZBTB17 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA589413 Lp-1 Miz1 Miz-1 Myc-interacting zinc finger protein 1 Myc-interacting Zn finger protein-1 mZ13 pHZ-67 Polyomavirus late initiator promoter-binding protein ZBTB17 Zfp100 Zfp-100 zinc finger and BTB domain containing 17 zinc finger and BTB domain-containing protein 17 Zinc finger protein 100 zinc finger protein 151 zinc finger protein 151 (pHZ-67) zinc finger protein 60 Zinc finger protein Z13 Znf151 ZNF60; LP-1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; Polyomavirus late initiator promoter-binding protein; Zfp-100; Zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; Zinc finger protein 151; zinc finger protein 151 (pHZ-67); Zinc finger protein 60; Zinc finger protein Z13
Gene Aliases: AA589413; Lp-1; MIZ-1; MIZ1; mZ13; pHZ-67; ZBTB17; Zfp100; ZNF151; ZNF60
UniProt ID: (Human) Q13105, (Mouse) Q60821
Entrez Gene ID: (Human) 7709, (Rat) 313666, (Mouse) 22642
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.