Abnova
Manufacturer Code:PAB20319
Catalog # 89-121-822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ZBTB17. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ZBTB17.
For Research Use Only
Protein Aliases: MIZ-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; pHZ-67; Zinc finger and BTB domain-containing protein 17; Zinc finger protein 151; zinc finger protein 151 (pHZ-67); Zinc finger protein 60; ZNF151; ZNF60
Gene Aliases: MIZ-1; MIZ1; pHZ-67; ZBTB17; ZNF151; ZNF60
UniProt ID: (Human) Q13105
Entrez Gene ID: (Human) 7709
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.