Abnova
Manufacturer Code:PAB8181
Catalog # 89-133-618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y334 of human ZBTB16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ZBTB16.
For Research Use Only
Protein Aliases: PLZF; promyelocytic leukaemia zinc finger; Promyelocytic leukemia zinc finger protein; Zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); Zinc finger protein PLZF; ZNF145
Gene Aliases: PLZF; ZBTB16; ZNF145
UniProt ID: (Human) Q05516
Entrez Gene ID: (Human) 7704
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.