Abnova
Manufacturer Code:PAB0318
Catalog # 89-114-136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 62-77 of human ZBTB10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: Zinc finger and BTB domain-containing protein 10; Zinc finger protein RIN ZF; zinc finger protein RINZF
Gene Aliases: RINZF; RINZFC; ZBTB10
UniProt ID: (Human) Q96DT7
Entrez Gene ID: (Human) 65986
Molecular Function: actin family cytoskeletal protein cytoskeletal protein hydrolase non-motor actin binding protein protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.