Proteintech
Manufacturer Code:229441AP150UL
Catalog # 50-173-3970
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZBTB10 Fusion Protein Ag19094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RINZF RINZFC ZBTB10 Zinc finger protein RIN ZF; Zinc finger and BTB domain-containing protein 10; Zinc finger protein RIN ZF; zinc finger protein RINZF
Gene Aliases: 4832414A18; AU021107; RINZF; RINZFC; ZBTB10
UniProt ID: (Human) Q96DT7
Entrez Gene ID: (Human) 65986, (Mouse) 229055
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.