Novus Biologicals
Manufacturer Code:NBP280056
Catalog # NB013184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Zippy-1 |
Immunogen | Recombinant mouse ZBP1/DLM-1/DAI (aa 1-411) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C20orf183 chromosome 20 open reading frame 183 DAI DLM1 DLM-1 DLM1dJ718J7.3 DNA-dependent activator of interferon regulatory factors DNA-dependent activator of IRFs Tumor stroma and activated macrophage protein DLM-1 ZBP1 Z-DNA binding protein 1 Z-DNA binding protein 1 Z-DNA-binding protein 1; DAI; DNA-dependent activator of IFN-regulatory factors; DNA-dependent activator of IRFs; Tumor stroma and activated macrophage protein DLM-1; Z-DNA-binding protein 1
Gene Aliases: C20orf183; DAI; DLM-1; DLM1; ZBP1
UniProt ID: (Human) Q9H171
Entrez Gene ID: (Human) 81030
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.