R&D Systems
Manufacturer Code:AF6309
Catalog # AF6309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ZBP1/DLM-1/DAI Ala2-Ile429 Accession # Q9H171 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf183 chromosome 20 open reading frame 183 DAI DLM1 DLM-1 DLM1dJ718J7.3 DNA-dependent activator of interferon regulatory factors DNA-dependent activator of IRFs Tumor stroma and activated macrophage protein DLM-1 ZBP1 Z-DNA binding protein 1 Z-DNA binding protein 1 Z-DNA-binding protein 1; DAI; DNA-dependent activator of IFN-regulatory factors; DNA-dependent activator of IRFs; Tumor stroma and activated macrophage protein DLM-1; Z-DNA-binding protein 1
Gene Aliases: C20orf183; DAI; DLM-1; DLM1; ZBP1
UniProt ID: (Human) Q9H171
Entrez Gene ID: (Human) 81030
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.