Invitrogen
Manufacturer Code:PA520455
Catalog # PIPA520455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide from near the carboxy terminus of human ZBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAI; DNA-dependent activator of IFN-regulatory factors; DNA-dependent activator of IRFs; RP4-718J7.1 C20orf183 DAI DLM-1 DLM1 DNA-dependent activator of IFN-regulatory factors DNA-dependent activator of IRFs DNA-dependent activator of interferon regulatory factors Z-DNA-binding protein 1 tumor stroma and activated macrophage protein DLM-1; Tumor stroma and activated macrophage protein DLM-1; Z-DNA-binding protein 1
Gene Aliases: 2010010H03Rik; C20orf183; DAI; DLM-1; DLM1; mZaDLM; ZBP1
UniProt ID: (Human) Q9H171, (Mouse) Q9QY24
Entrez Gene ID: (Human) 81030, (Mouse) 58203, (Rat) 171091
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.