Invitrogen
Manufacturer Code:PA561747
Catalog # PIPA561747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZBED5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Transposon-derived Buster1 transposase-like protein; Zinc finger BED domain-containing protein 5; Zinc finger BED-type containing 5 ZBED5 Buster1; zinc finger, BED-type containing 5
Gene Aliases: Buster1; ZBED5
UniProt ID: (Human) Q49AG3
Entrez Gene ID: (Human) 58486
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.