Novus Biologicals
Manufacturer Code:NBP288603
Catalog # NB036225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human ZBED5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Transposon-derived Buster1 transposase-like protein; Zinc finger BED domain-containing protein 5; zinc finger BED domain-containing protein 5 zinc finger BED-type containing 5; zinc finger, BED-type containing 5
Gene Aliases: Buster1; ZBED5
UniProt ID: (Human) Q49AG3
Entrez Gene ID: (Human) 58486
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.