Novus Biologicals
Manufacturer Code:NBP177348
Catalog # NBP177348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human ZBED3. The immunogen is located within the last 50 amino acids of ZBED3. Amino Acid Squence: APPPLKDDPEGDRDG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610005H11Rik; Axin-interacting protein; BED domain containing 3 MGC154352610005H11Rik zinc finger BED-type containing 3; Zinc finger BED domain-containing protein 3; zinc finger, BED domain containing 3; zinc finger, BED-type containing 3
Gene Aliases: ZBED3
UniProt ID: (Human) Q96IU2
Entrez Gene ID: (Human) 84327
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.