Novus Biologicals
Manufacturer Code:NBP2445050.2MG
Catalog # NBP24450502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ZAP70/528 + 2F3.2 |
Immunogen | Recombinant full-length human ZAP70 protein (ZAP70/528) Recombinant ZAP-70 protein including residues 1-254 and encompassing SH2 domains of human ZAP70 (2F3.2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa zeta-associated protein; 70 kDa zeta-associated protein EC 2.7.10 EC 2.7.10.2 SRKFLJ17679 STDFLJ17670 Syk-related tyrosine kinase tyrosine-protein kinase ZAP-70 TZK ZAP-70 zeta-chain (TCR) associated protein kinase (70 kD) zeta-chain (TCR) associated protein kinase 70kDa zeta-chain associated protein kinase 70kD; 70 kDa zeta-chain associated protein; Syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Aliases: ADMIO2; IMD48; SRK; STCD; STD; TZK; ZAP-70; ZAP70
UniProt ID: (Human) P43403
Entrez Gene ID: (Human) 7535
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.