Abnova
Manufacturer Code:PAB18235
Catalog # 89-119-805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ZAP70. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ZAP70.
For Research Use Only
Protein Aliases: 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; FLJ17670; FLJ17679; SRK; STD; Syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; TZK; ZAP-70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Aliases: ADMIO2; IMD48; SRK; STCD; STD; TZK; ZAP-70; ZAP70
UniProt ID: (Human) P43403
Entrez Gene ID: (Human) 7535
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.