Proteintech
Manufacturer Code:146791AP150UL
Catalog # 50-172-9635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZADH2 Fusion Protein Ag6390 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15-oxoprostaglandin 13-reductase; PRG-3; Prostaglandin reductase 3; Prostaglandin reductase-3; PTGR-3; ZADH2; Zinc-binding alcohol dehydrogenase domain-containing protein 2
Gene Aliases: C530046K17Rik; PTGR3; Pthr3; ZADH2
UniProt ID: (Human) Q8N4Q0, (Mouse) Q8BGC4
Entrez Gene ID: (Human) 284273, (Rat) 291403, (Mouse) 225791
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.