Invitrogen
Manufacturer Code:PA577765
Catalog # PIPA577765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CNFELLHFPRDHSN corresponding to amino acid residues 157-170 of human zinc-activated channel (ZAC) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ligand-gated ion channel subunit; Ligand-gated ion channel zinc-activated 1; ligand-gated ion channel, zinc activated 1; Ligand-gated ion-channel receptor L2; ZACN; zinc activated ligand-gated ion channel 1; Zinc-activated ligand-gated ion channel
Gene Aliases: L2; LGICZ; LGICZ1; ZAC; ZAC1; ZACN
UniProt ID: (Human) Q401N2
Entrez Gene ID: (Human) 353174
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.